accessibility menu, dialog, popup

UserName

ClClONON

CAS RN 84-58-2

IUPAC Name: 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
Synonyms: 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
Molecular Weight (g/mol): 227
Molecular Formula: C8Cl2N2O2
InChi Key: HZNVUJQVZSTENZ-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)