accessibility menu, dialog, popup

UserName

CH3H3CBOHHO

CAS RN 84110-40-7

IUPAC Name: (2-methylpropyl)boronic acid
Synonyms: 2-methylpropylboronic acid
Molecular Weight (g/mol): 101.94
Molecular Formula: C4H11BO2
InChi Key: ZAZPDOYUCVFPOI-UHFFFAOYSA-N
SMILES: CC(C)CB(O)O
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)