accessibility menu, dialog, popup

UserName

H3CONNOF

CAS RN 84163-10-0

IUPAC Name: 1-[4-(5-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethan-1-one
Synonyms: 1-[4-(5-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethanone
Molecular Weight (g/mol): 262.28
Molecular Formula: C14H15FN2O2
InChi Key: PZNSVLCWHLHDLK-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC(CC1)C1=NOC2=CC=C(F)C=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results