accessibility menu, dialog, popup

UserName

CAS RN 842-07-9

ONHN(Z)

CAS RN 842-07-9

IUPAC Name: (1Z)-1-(2-phenylhydrazin-1-ylidene)-1,2-dihydronaphthalen-2-one
Synonyms: CI solvent yellow 14
Molecular Weight (g/mol): 248.29
Molecular Formula: C16H12N2O
InChi Key: ZONYAPYTDIVJGG-VLGSPTGOSA-N
SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=C1
Color 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)