accessibility menu, dialog, popup

UserName

CH3NNOOH3CH3CCH3CH3O

CAS RN 844891-13-0

IUPAC Name: tert-butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate
Synonyms: tert-butyl 4-formyl-3,5-dimethylpyrazole-1-carboxylate
Molecular Weight (g/mol): 224.26
Molecular Formula: C11H16N2O3
InChi Key: AZJWODKWFMBMCM-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)OC(C)(C)C)C(C)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results