accessibility menu, dialog, popup

UserName

H3CNCH3NNOONa(E/Z)

CAS RN 845-46-5

IUPAC Name: sodium 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Synonyms: sodium 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular Weight (g/mol): 291.29
Molecular Formula: C15H14N3NaO2
InChi Key: OSCKRHPYZNTEIO-UHFFFAOYSA-M
SMILES: [Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C([O-])=O
Grade 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)