CAS RN 848243-23-2
IUPAC Name:
2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms:
2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol):
249.12
Molecular Formula:
C13H20BNO3
InChi Key:
GRXNEZGBXVIORK-UHFFFAOYSA-N
SMILES:
CCOC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
2-Ethoxypyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™
CAS: 848243-23-2 Molecular Formula: C13H20BNO3 Molecular Weight (g/mol): 249.12 MDL Number: MFCD07366721 InChI Key: GRXNEZGBXVIORK-UHFFFAOYSA-N Synonym: 2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-ethoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine,2-ethoxypyridine-3-boronic acid pinacol ester,2-ethoxypyridin-3-ylboronic acid pinacol ester,2-ethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,pubchem16583,2-ethoxypyridin-3-yl boronic acid pinacol ester PubChem CID: 17750191 IUPAC Name: 2-ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine SMILES: CCOC1=C(C=CC=N1)B1OC(C)(C)C(C)(C)O1