accessibility menu, dialog, popup

UserName

CAS RN 849066-63-3

CH3NONOCH3OHO

CAS RN 849066-63-3

IUPAC Name: 3-methyl-5-(5-methyl-1,2-oxazol-3-yl)-1,2-oxazole-4-carboxylic acid
Synonyms: 3-methyl-5-(5-methyl-1,2-oxazol-3-yl)-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 208.17
Molecular Formula: C9H8N2O4
InChi Key: ASBPDKJMOPNMLF-UHFFFAOYSA-N
SMILES: CC1=CC(=NO1)C1=C(C(O)=O)C(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results