accessibility menu, dialog, popup

UserName

CAS RN 84940-46-5

NFFFFFF

CAS RN 84940-46-5

IUPAC Name: 2,3,6-trifluoro-4-(trifluoromethyl)pyridine
Synonyms: 2,3,6-trifluoro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 201.07
Molecular Formula: C6HF6N
InChi Key: DODFHNWFNCGXOW-UHFFFAOYSA-N
SMILES: FC1=NC(F)=C(F)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results