CAS RN 84954-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Rosavin, TRC
CAS: 84954-92-7 Molecular Formula: C20 H28 O10 Molecular Weight (g/mol): 428.43 Synonym: Rosavin, Rosavidin, (2E)-3-phenyl-2-propenyl 6-O-alpha-L-arabinopyranosyl-beta-D-Glucopyranoside IUPAC Name: (2R,3R,4S,5S,6R)-2-[(E)-3-phenylprop-2-enoxy]-6-[[(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl]oxane-3,4,5-triol SMILES: C(O[C@H]1[C@H](O)[C@@H](O)[C@@H](O)CO1)[C@H]2O[C@@H](OC/C=C/C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O