accessibility menu, dialog, popup

UserName

CAS RN 84978-67-6

ONHNHO

CAS RN 84978-67-6

IUPAC Name: [3-(furan-2-yl)-1H-pyrazol-5-yl]methanol
Synonyms: [5-(furan-2-yl)-2H-pyrazol-3-yl]methanol
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: NBADTAIDQSLLHA-UHFFFAOYSA-N
SMILES: OCC1=CC(=NN1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results