accessibility menu, dialog, popup

UserName

CH3SNNHCH3

CAS RN 850375-02-9

IUPAC Name: methyl[(4-methyl-2-phenyl-1,3-thiazol-5-yl)methyl]amine
Synonyms: methyl[(4-methyl-2-phenyl-1,3-thiazol-5-yl)methyl]amine
Molecular Weight (g/mol): 218.32
Molecular Formula: C12H14N2S
InChi Key: GFAGRBRYZWAUSV-UHFFFAOYSA-N
SMILES: CNCC1=C(C)N=C(S1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results