accessibility menu, dialog, popup

UserName

CAS RN 850568-10-4

IUPAC Name: (3-carbamothioylphenyl)boronic acid
Synonyms: 3-carbamothioylphenylboronic acid
Molecular Weight (g/mol): 181.02
Molecular Formula: C7H8BNO2S
InChi Key: ZPKLUYJBCAHWIW-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results