accessibility menu, dialog, popup

UserName

CAS RN 850568-59-1

IUPAC Name: (4-ethoxy-3,5-dimethylphenyl)boronic acid
Synonyms: 4-ethoxy-3,5-dimethylphenylboronic acid
Molecular Weight (g/mol): 194.04
Molecular Formula: C10H15BO3
InChi Key: FMJASQDMWVBTOJ-UHFFFAOYSA-N
SMILES: CCOC1=C(C)C=C(C=C1C)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results