accessibility menu, dialog, popup

UserName

BNFFFK

CAS RN 850623-46-0

IUPAC Name: potassium (3-cyanophenyl)trifluoroboranuide
Synonyms: potassium (3-cyanophenyl)trifluoroboranuide
Molecular Weight (g/mol): 209.02
Molecular Formula: C7H4BF3KN
InChi Key: STQLGUGARUCKLI-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC(=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results