accessibility menu, dialog, popup

UserName

BOFFFK

CAS RN 850623-47-1

IUPAC Name: potassium [4-(benzyloxy)phenyl]trifluoroboranuide
Synonyms: potassium [4-(benzyloxy)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 290.13
Molecular Formula: C13H11BF3KO
InChi Key: PMDBNNXIBAIJMQ-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results