accessibility menu, dialog, popup

UserName

OOO

CAS RN 85064-61-5

IUPAC Name: 2-(oxan-4-yl)acetate
Synonyms: 2-(oxan-4-yl)acetate
Molecular Weight (g/mol): 143.16
Molecular Formula: C7H11O3
InChi Key: PBXYNWPYMVWJAH-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results