accessibility menu, dialog, popup

UserName

CAS RN 852227-90-8

BOOH3CCH3H3CH3CN

CAS RN 852227-90-8

IUPAC Name: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
Synonyms: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
Molecular Weight (g/mol): 273.18
Molecular Formula: C16H24BNO2
InChi Key: DWJNNJSONWFVBT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)