accessibility menu, dialog, popup

UserName

H3CNOH

CAS RN 854778-61-3

IUPAC Name: (1-methyl-1H-indol-7-yl)methanol
Synonyms: (1-methylindol-7-yl)methanol
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: XWTPJITYDAXWKC-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC(CO)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results