accessibility menu, dialog, popup

UserName

CAS RN 857283-56-8

IUPAC Name: 5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
Synonyms: 5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
Molecular Weight (g/mol): 257.69
Molecular Formula: C10H8ClNO3S
InChi Key: GXQBTNFENYVPFM-UHFFFAOYSA-N
SMILES: CC1=C(C(=NO1)C1=CC=CC=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results