accessibility menu, dialog, popup

UserName

NNCO

CAS RN 857283-59-1

IUPAC Name: 1-(3-isocyanatophenyl)-1H-pyrrole
Synonyms: 1-(3-isocyanatophenyl)pyrrole
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: PUPFPJGLDBUOOI-UHFFFAOYSA-N
SMILES: O=C=NC1=CC(=CC=C1)N1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results