accessibility menu, dialog, popup

UserName

CAS RN 857283-66-0

CH3SNNOOH3CCH3H3COH

CAS RN 857283-66-0

IUPAC Name: tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate
Molecular Weight (g/mol): 312.43
Molecular Formula: C15H24N2O3S
InChi Key: YNLQKGMVENCCLE-UHFFFAOYSA-N
SMILES: CC1=C(CO)SC(=N1)C1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results