accessibility menu, dialog, popup

UserName

NH2SNH

CAS RN 86-88-4

IUPAC Name: (naphthalen-1-yl)thiourea
Synonyms: α-naphthylthiourea antu (iso) N-1-napthylthiourea
Molecular Weight (g/mol): 202.28
Molecular Formula: C11H10N2S
InChi Key: PIVQQUNOTICCSA-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)