CAS RN 86008-21-1
CAS RN 86008-21-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-O-Octadecyl-2-O-benzyl-rac-glycerol, TRC
CAS: 86008-21-1 Molecular Formula: C28H50O3 Molecular Weight (g/mol): 434.69 Synonym: 3-(Octadecyloxy)-2-(phenylmethoxy)-1-propanol,2-(Benzyloxy)-3-(octadecyloxy)propan-1-ol IUPAC Name: 3-octadecoxy-2-phenylmethoxypropan-1-ol SMILES: CCCCCCCCCCCCCCCCCCOCC(CO)OCc1ccccc1