accessibility menu, dialog, popup

UserName

CAS RN 860296-24-8

NFClOHO

CAS RN 860296-24-8

IUPAC Name: 6-chloro-5-fluoropyridine-2-carboxylic acid
Synonyms: 6-chloro-5-fluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: JHFKXHUKNYFWJM-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(Cl)=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results