accessibility menu, dialog, popup

UserName

CH3NNOO

CAS RN 861119-08-6

IUPAC Name: 2-{5-methyl-octahydropyrrolo[3,4-c]pyrrol-2-yl}-9H-xanthen-9-one
Synonyms: 2-{5-methyl-hexahydropyrrolo[3,4-c]pyrrol-2-yl}xanthen-9-one
Molecular Weight (g/mol): 320.39
Molecular Formula: C20H20N2O2
InChi Key: WUJHMDPPANTBTQ-UHFFFAOYNA-N
SMILES: CN1CC2CN(CC2C1)C1=CC=C2OC3=CC=CC=C3C(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results