CAS RN 861891-50-1
IUPAC Name:
1-methyl-3-(1-methyl-1H-indol-3-yl)-4-(pentylamino)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms:
1-methyl-3-(1-methylindol-3-yl)-4-(pentylamino)pyrrole-2,5-dione
Molecular Weight (g/mol):
325.41
Molecular Formula:
C19H23N3O2
InChi Key:
SGLOMINNEBLJFF-UHFFFAOYSA-N
SMILES:
CCCCCNC1=C(C(=O)N(C)C1=O)C1=CN(C)C2=CC=CC=C12