CAS RN 86299-47-0
IUPAC Name:
(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-({[1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl]oxy}imino)acetate
Synonyms:
(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-({[1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl]oxy}imino)acetate
Molecular Weight (g/mol):
328.36
Molecular Formula:
C13H18N3O5S
InChi Key:
FNRZBOJFRDVEOG-LZYBPNLTSA-M
SMILES:
CC(C)(C)OC(=O)C(C)(C)O\N=C(\C([O-])=O)C1=CSC(N)=N1
1
–
3
of
3
results
1
(Z)-2-(2-Aminothiazol-4-yl)-2-[(1-tert-butoxycarbonyl-1-methylethoxy)imino]acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(Z)-2-(2-Aminothiazol-4-yl)-2-[(1-tert-butoxycarbonyl-1-methylethoxy)imino]acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(Z)-2-(2-Aminothiazol-4-yl)-2-[(1-tert-butoxycarbonyl-1-methylethoxy)imino]acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.