CAS RN 86303-22-2
IUPAC Name:
2,3,4,5,6-pentahydroxy-N-(3-{N-[3-(2,3,4,5,6-pentahydroxyhexanamido)propyl]-4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido}propyl)hexanamide
Synonyms:
2,3,4,5,6-pentahydroxy-N-(3-{N-[3-(2,3,4,5,6-pentahydroxyhexanamido)propyl]-4-{4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido}propyl)hexanamide
Molecular Weight (g/mol):
878.07
Molecular Formula:
C42H75N3O16
InChi Key:
ZWEVPYNPHSPIFU-UHFFFAOYNA-N
SMILES:
CC(CCC(=O)N(CCCNC(=O)C(O)C(O)C(O)C(O)CO)CCCNC(=O)C(O)C(O)C(O)C(O)CO)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C
1
–
4
of
4
results
2
N,N-Bis[3-D-gluconamidopropyl]cholamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N,N-Bis[3-D-gluconamidopropyl]cholamide (Ultra Pure), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N,N-Bis[3-D-gluconamidopropyl]cholamide (Ultra Pure), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.