CAS RN 86311-35-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Amino-2-methylpropane-1-sulfonic acid, TRC
CAS: 86311-35-5 Molecular Formula: C4 H11 N O3 S Molecular Weight (g/mol): 153.2 IUPAC Name: 2-amino-2-methylpropane-1-sulfonic acid SMILES: CC(C)(N)CS(=O)(=O)O