accessibility menu, dialog, popup

UserName

ONNO

CAS RN 864068-87-1

IUPAC Name: 2-(morpholin-4-yl)pyridine-4-carbaldehyde
Synonyms: 2-(morpholin-4-yl)pyridine-4-carbaldehyde
Molecular Weight (g/mol): 192.22
Molecular Formula: C10H12N2O2
InChi Key: VMFCHBZYIHUMQE-UHFFFAOYSA-N
SMILES: O=CC1=CC(=NC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results