accessibility menu, dialog, popup

UserName

CH3NNOH

CAS RN 864068-97-3

IUPAC Name: (1-methyl-3-phenyl-1H-pyrazol-5-yl)methanol
Synonyms: (2-methyl-5-phenylpyrazol-3-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: GRHZYDGKXPVLEH-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1CO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results