CAS RN 864754-13-2
IUPAC Name:
tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
Synonyms:
tert-butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
Molecular Weight (g/mol):
416.33
Molecular Formula:
C22H33BN2O5
InChi Key:
QPYOLSJFOCIOHK-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1CCN(CC1)C(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
2
of
2
results
1
4-(4-Boc-piperazine-1-carbonyl)phenylboronic acid pinacol ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(4-Boc-piperazine-1-carbonyl)phenylboronic acid pinacol ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.