accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CNNNO

CAS RN 864754-18-7

IUPAC Name: 4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethyl}morpholine
Synonyms: 4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]ethyl}morpholine
Molecular Weight (g/mol): 307.2
Molecular Formula: C15H26BN3O3
InChi Key: FBAPTUAEBQMVEY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(CCN2CCOCC2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results