CAS RN 865758-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-des(piperidin-3R-amino)-1-chloro-Alogliptin, TRC
CAS: 865758-96-9 Molecular Formula: C13H10ClN3O2 Molecular Weight (g/mol): 275.69 Synonym: 2-[(6-Chloro-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl)methyl]-benzonitrile,Alogliptin 6-Chloro Impurity IUPAC Name: 2-[(6-chloro-3-methyl-2,4-dioxopyrimidin-1-yl)methyl]benzonitrile SMILES: CN1C(=O)C=C(Cl)N(Cc2ccccc2C#N)C1=O