accessibility menu, dialog, popup

UserName

ClHSnHClCH3H3C

CAS RN 866-55-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 249.75
Molecular Formula: C4H12Cl2Sn
InChi Key: RFGFYUBDZRGQMZ-UHFFFAOYSA-N
SMILES: Cl.Cl.CC[SnH2]CC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)