accessibility menu, dialog, popup

UserName

NNHNNClHClHClHCl

CAS RN 866021-03-6

IUPAC Name: 1-(4-chlorophenyl)-4-({1H-pyrrolo[2,3-b]pyridin-3-yl}methyl)piperazine trihydrochloride
Synonyms: 1-(4-chlorophenyl)-4-{1H-pyrrolo[2,3-b]pyridin-3-ylmethyl}piperazine trihydrochloride
Molecular Weight (g/mol): 436.2
Molecular Formula: C18H22Cl4N4
InChi Key: KJSOYZLYCFYXFC-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.ClC1=CC=C(C=C1)N1CCN(CC2=CNC3=NC=CC=C23)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results