accessibility menu, dialog, popup

UserName

OOPBr

CAS RN 86608-70-0

IUPAC Name: [2-(1,3-dioxolan-2-yl)ethyl]triphenylphosphanium bromide
Synonyms: [2-(1,3-dioxolan-2-yl)ethyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 443.32
Molecular Formula: C23H24BrO2P
InChi Key: ZCJKBPSRKLHANV-UHFFFAOYSA-M
SMILES: [Br-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1OCCO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)