accessibility menu, dialog, popup

UserName

HONHONNNHCl

CAS RN 866396-70-5

IUPAC Name: 1-(2-{[1-(hydroxymethyl)cyclopentyl]amino}acetyl)pyrrolidine-2,5-dicarbonitrile hydrochloride
Synonyms: 1-(2-{[1-(hydroxymethyl)cyclopentyl]amino}acetyl)pyrrolidine-2,5-dicarbonitrile hydrochloride
Molecular Weight (g/mol): 312.8
Molecular Formula: C14H21ClN4O2
InChi Key: RHCWAHDCHQTNBR-UHFFFAOYNA-N
SMILES: Cl.OCC1(CCCC1)NCC(=O)N1C(CCC1C#N)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results