accessibility menu, dialog, popup

UserName

ClHNNSNH2HClHCl(E/Z)

CAS RN 866914-87-6

IUPAC Name: N'-[4-(2-{[(3-chlorophenyl)methyl]amino}ethyl)phenyl]thiophene-2-carboximidamide dihydrochloride
Synonyms: N'-[4-(2-{[(3-chlorophenyl)methyl]amino}ethyl)phenyl]thiophene-2-carboximidamide dihydrochloride
Molecular Weight (g/mol): 442.83
Molecular Formula: C20H22Cl3N3S
InChi Key: SOXBYIKSUDXMIE-UHFFFAOYSA-N
SMILES: Cl.Cl.NC(=NC1=CC=C(CCNCC2=CC(Cl)=CC=C2)C=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results