CAS RN 866926-81-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Scopolamine Sulfate, TRC
CAS: 866926-81-0 Molecular Formula: C17H21NO7S Molecular Weight (g/mol): 383.42 Synonym: α-[(Sulfooxy)methyl]benzeneacetic Acid C-[(1α,2α,4α,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl] Ester SMILES: CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](COS(=O)(=O)O)c4ccccc4