accessibility menu, dialog, popup

UserName

H3CSNH2HNH3CSNH2HNOHSOHOO

CAS RN 867-44-7

IUPAC Name: bis((methylsulfanyl)methanimidamide); sulfuric acid
Synonyms: bis(methylsulfanylmethanimidamide); sulfuric acid
Molecular Weight (g/mol): 278.36
Molecular Formula: C4H14N4O4S3
InChi Key: BZZXQZOBAUXLHZ-UHFFFAOYSA-N
SMILES: CSC(N)=N.CSC(N)=N.OS(O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)