CAS RN 86770-67-4
IUPAC Name:
2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethan-1-ol
Synonyms:
2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethanol
Molecular Weight (g/mol):
219.24
Molecular Formula:
C8H17N3O4
InChi Key:
MBQYGQMGPFNSAP-UHFFFAOYSA-N
SMILES:
OCCOCCOCCOCCN=[N+]=[N-]
1
–
2
of
2
results
1
PEG4-azide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
PEG4-azide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.