CAS RN 86786-39-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Fluoro 2-Deoxy-2-fluoro-3,4,6-tri-O-acetyl-D-glucose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fluoro 2-Deoxy-2-fluoro-3,4,6-tri-O-acetyl-D-glucose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Fluoro 2-Deoxy-2-fluoro-3,4,6-tri-O-acetyl-D-glucose, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.