accessibility menu, dialog, popup

UserName

H3COOCH3ONOOHNOO

CAS RN 868224-64-0

IUPAC Name: ethyl 2-[(2-{[(2,3-dihydro-1,4-benzodioxin-6-yl)carbamoyl]methyl}-1-oxo-1,2-dihydroisoquinolin-5-yl)oxy]propanoate
Synonyms: ethyl 2-[(2-{[(2,3-dihydro-1,4-benzodioxin-6-yl)carbamoyl]methyl}-1-oxoisoquinolin-5-yl)oxy]propanoate
Molecular Weight (g/mol): 452.46
Molecular Formula: C24H24N2O7
InChi Key: KWMBIIQCLUIHDI-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)OC1=C2C=CN(CC(=O)NC3=CC4=C(OCCO4)C=C3)C(=O)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results