accessibility menu, dialog, popup

UserName

CH3H3CCH3OOONCH3OOHHCl

CAS RN 868265-28-5

IUPAC Name: 2-({2-[4-(2H-1,3-benzodioxol-5-yl)-2-tert-butylphenoxy]ethyl}(methyl)amino)acetic acid hydrochloride
Synonyms: ({2-[4-(2H-1,3-benzodioxol-5-yl)-2-tert-butylphenoxy]ethyl}(methyl)amino)acetic acid hydrochloride
Molecular Weight (g/mol): 421.92
Molecular Formula: C22H28ClNO5
InChi Key: ZQVOAGQZHDAFRM-UHFFFAOYSA-N
SMILES: Cl.CN(CCOC1=C(C=C(C=C1)C1=CC2=C(OCO2)C=C1)C(C)(C)C)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results