accessibility menu, dialog, popup

UserName

NH2H2NH2NNH2HClHClHClHCl

CAS RN 868272-85-9

IUPAC Name: [1,1'-biphenyl]-3,3',4,4'-tetramine hydrate
Synonyms: 3,3'-diaminobenzidine hydrate
Molecular Weight (g/mol): 232.29
Molecular Formula: C12H16N4O
InChi Key: WWIHRTPUMJFLTN-UHFFFAOYSA-N
SMILES: O.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)