accessibility menu, dialog, popup

UserName

CAS RN 86847-64-5

CH3H3CCH3OHNNO

CAS RN 86847-64-5

IUPAC Name: N-(3-formylpyridin-2-yl)-2,2-dimethylpropanamide
Synonyms: N-(3-formylpyridin-2-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: ANABHCSYKASRRW-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=NC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results