accessibility menu, dialog, popup

UserName

NNOH2N

CAS RN 868696-42-8

IUPAC Name: 5-cyclopropyl-1,2,4-oxadiazol-3-amine
Synonyms: 5-cyclopropyl-1,2,4-oxadiazol-3-amine
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: OHSZRALIDGBNES-UHFFFAOYSA-N
SMILES: NC1=NOC(=N1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results