accessibility menu, dialog, popup

UserName

CH3OHNCH3

CAS RN 869901-18-8

IUPAC Name: methyl[(5-methyl-2-phenylfuran-3-yl)methyl]amine
Synonyms: methyl[(5-methyl-2-phenylfuran-3-yl)methyl]amine
Molecular Weight (g/mol): 201.27
Molecular Formula: C13H15NO
InChi Key: UOPUGXOLGTYISK-UHFFFAOYSA-N
SMILES: CNCC1=C(OC(C)=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results